Gene Transcripts

Gene Symbol
NAB1
Entrez Gene ID
4664
Gene Name
NGFI-A binding protein 1
Gene Aliases
Chromosome Location
Chr. 2: 190648860 - 190692766 on Build GRCh38
UniGene ID
Hs.107474
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321312.1 NP_001308241.1 5 1373
NM_001321313.1 NP_001308242.1 5 1159
NM_001321314.1 NP_001308243.1 4 1082
NM_005966.3 NP_005957.2 7 1635
XM_005246582.1 XP_005246639.1 6 1308
XM_005246583.1 XP_005246640.1 6 1422
XM_005246586.1 XP_005246643.1 5 1115
XM_011511219.2 XP_011509521.1 6 1277
XM_017004170.1 XP_016859659.1 6 1318
XM_017004171.1 XP_016859660.1 5 1488
XM_017004172.1 XP_016859661.1 5 1260
XM_017004173.1 XP_016859662.1 6 1343
XM_017004174.1 XP_016859663.1 5 1159
XM_017004176.1 XP_016859665.1 5 1227
GenBank mRNA AF045451.1 - 4 1082
AK292446.1 - 5 1173
AK313542.1 - 4 1180
BC035724.1 - 7 1524 5721972
U47007.1 - 4 1082

View Full Product Details