Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040092.2 | NP_001035181.1 | 22 | 2202 | |
NM_001130863.2 | NP_001124335.1 | 23 | 2277 | ||
NM_006209.4 | NP_006200.3 | 23 | 2358 | ||
NR_045555.1 | 11 | 1107 | |||
XM_006716584.1 | XP_006716647.1 | 24 | 2411 | ||
XM_006716585.1 | XP_006716648.1 | 24 | 2399 | ||
XM_006716587.1 | XP_006716650.1 | 23 | 2324 | ||
XM_017013571.1 | XP_016869060.1 | 23 | 2311 | ||
XM_017013572.1 | XP_016869061.1 | 23 | 2243 | ||
XM_017013573.1 | XP_016869062.1 | 23 | 2299 | ||
XM_017013574.1 | XP_016869063.1 | 22 | 2236 | ||
XM_017013575.1 | XP_016869064.1 | 22 | 2168 | ||
GenBank mRNA | AK124910.1 | - | 4 | 2737 | |
AK130313.1 | - | 11 | 1107 | ||
AK296027.1 | - | 8 | 818 | ||
AK315253.1 | - | 22 | 2175 | ||
AL544867.3 | - | 5 | 504 | ||
BC034961.2 | - | 22 | 2175 | 4828484 | |
EU131011.2 | - | 23 | 2191 | ||
L35594.1 | - | 23 | 2321 |