Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000950.2 | NP_000941.1 | 4 | 353 | |
NM_001142395.1 | NP_001135867.1 | 3 | 262 | ||
NM_001173486.1 | NP_001166957.1 | 3 | 262 | ||
NM_001173489.1 | NP_001166960.1 | 4 | 372 | ||
NM_001173490.1 | NP_001166961.1 | 3 | 281 | ||
GenBank mRNA | AF009242.1 | - | 4 | 265 | |
AK312904.1 | - | 3 | 201 | ||
BC030786.2 | - | 3 | 245 | ||
BC060833.1 | - | 3 | 201 | 30348133 | |
DA245329.1 | - | 4 | 339 | ||
DA411839.1 | - | 4 | 311 | ||
DA417581.1 | - | 3 | 281 | ||
DB286340.1 | - | 3 | 264 |