Gene Transcripts
- Gene Symbol
- RAD23A
- Entrez Gene ID
- 5886
- Gene Name
- RAD23 homolog A, nucleotide excision repair protein
- Gene Aliases
- HHR23A, HR23A
- Chromosome Location
- Chr. 19: 12945814 - 12953643 on Build GRCh38
- UniGene ID
- Hs.643267
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001270362.1 |
NP_001257291.1
|
5, 6 |
727 |
|
NM_001270363.1 |
NP_001257292.1
|
5, 6 |
727 |
|
NM_005053.3 |
NP_005044.1
|
5, 6 |
727 |
|
NR_072976.1 |
|
4, 5 |
671 |
|
GenBank mRNA |
AB209713.1 |
- |
5, 6 |
650 |
|
AK289908.1 |
- |
5, 6 |
664 |
|
AK293219.1 |
- |
4, 5 |
630 |
|
BC014026.2 |
- |
5, 6 |
653 |
4300551 |
BC088364.1 |
- |
5, 6 |
656 |
30389208 |
BX448989.2 |
- |
5, 6 |
648 |
|
D21235.1 |
- |
5, 6 |
628 |
|
M77024.1 |
- |
1, 2 |
40 |
|
View Full Product Details