Gene Transcripts

Gene Symbol
RBBP7
Entrez Gene ID
5931
Gene Name
RB binding protein 7, chromatin remodeling factor
Gene Aliases
RbAp46
Chromosome Location
Chr. X: 16844650 - 16870411 on Build GRCh38
UniGene ID
Hs.495755
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198719.1 NP_001185648.1 12 1609
NM_002893.3 NP_002884.1 12 1621
GenBank mRNA AK091911.1 - 12 1606
AK127332.1 - 3 3634
AK291567.1 - 12 1365
BC114500.1 - 12 1519 40083852
BC114501.1 - 12 1519 40083851
BQ775345.1 - 1 406
U35143.1 - 12 1524
X72841.1 - 12 1548

View Full Product Details