Gene Transcripts

Gene Symbol
RNASE6
Entrez Gene ID
6039
Gene Name
ribonuclease A family member k6
Gene Aliases
RAD1, RNS6, RNasek6
Chromosome Location
Chr. 14: 20780956 - 20782467 on Build GRCh38
UniGene ID
Hs.23262
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005615.4 NP_005606.1 2 394
XM_017021566.1 XP_016877055.1 3 281
XM_017021567.1 XP_016877056.1 3 294
GenBank mRNA AK313580.1 - 2 377
BC020848.1 - 2 394 4732021
CR456965.1 - 1 101

View Full Product Details