Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003363.3 | NP_003354.2 | 19 | 2575 | |
NM_199443.2 | NP_955475.1 | 18 | 2434 | ||
GenBank mRNA | AF017305.1 | - | 19 | 2499 | |
AF017306.1 | - | 18 | 2355 | ||
AK127055.1 | - | 13 | 3011 | ||
AK222725.1 | - | 18 | 2364 | ||
AK223292.1 | - | 19 | 2505 | ||
AK291795.1 | - | 19 | 2520 | ||
BC125130.1 | - | 19 | 2546 | 40119090 | |
BC125131.1 | - | 19 | 2543 | 40119091 | |
U20657.1 | - | 19 | 2499 |