Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001077516.1 | NP_001070984.1 | 2 | 264 | |
NM_006979.2 | NP_008910.2 | 1 | 567 | ||
GenBank mRNA | AK225282.1 | - | 1 | 491 | |
AK225682.1 | - | 1 | 446 | ||
BC000645.2 | - | 2 | 202 | 3345970 | |
CB107807.1 | - | 2 | 264 | ||
D82060.1 | - | 1 | 444 | ||
DA573310.1 | - | 2 | 264 | ||
DR158641.1 | - | 1 | 471 | ||
M58660.1 | - | 1 | 126 |