Gene Transcripts

Gene Symbol
PIK3R3
Entrez Gene ID
8503
Gene Name
phosphoinositide-3-kinase regulatory subunit 3
Gene Aliases
p55, p55-GAMMA, p55PIK
Chromosome Location
Chr. 1: 46040140 - 46176495 on Build GRCh38
UniGene ID
Hs.655387
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114172.1 NP_001107644.1 10 1387
NM_001303427.1 NP_001290356.1 9 1292
NM_001303428.1 NP_001290357.1 10 1231
NM_001303429.1 NP_001290358.1 8 1709
NM_003629.3 NP_003620.3 9 1886
XM_017002614.1 XP_016858103.1 10 1264
GenBank mRNA AF028785.1 - 8 1644
AK302049.1 - 9 1285
AK304713.1 - 4 576
AK313726.1 - 10 1399
BC021622.1 - 9 1820 3910221
BX537388.1 - 10 1231
CR749649.1 - 9 1807
D88532.1 - 9 1550

View Full Product Details