Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001161441.1 | NP_001154913.1 | 8 | 1200 | |
NM_001161442.1 | NP_001154914.1 | 8 | 1139 | ||
NM_001161443.1 | NP_001154915.1 | 7 | 1049 | ||
NM_001161444.1 | NP_001154916.1 | 8 | 1170 | ||
NM_003975.3 | NP_003966.2 | 8 | 1170 | ||
XM_006711615.2 | XP_006711678.1 | 8 | 1188 | ||
XM_017002762.1 | XP_016858251.1 | 8 | 1217 | ||
XM_017002763.1 | XP_016858252.1 | 8 | 1208 | ||
XM_017002764.1 | XP_016858253.1 | 8 | 1155 | ||
XM_017002765.1 | XP_016858254.1 | 8 | 1168 | ||
XM_017002766.1 | XP_016858255.1 | 8 | 1158 | ||
XM_017002767.1 | XP_016858256.1 | 7 | 935 | ||
GenBank mRNA | AF051325.1 | - | 8 | 1092 | |
AF097744.1 | - | 8 | 1069 | ||
AJ000553.1 | - | 8 | 1116 | ||
AK222737.1 | - | 8 | 1093 | ||
AK313706.1 | - | 8 | 1164 | ||
AY763098.1 | - | 7 | 1049 | ||
AY763099.1 | - | 8 | 1146 | ||
AY763100.1 | - | 8 | 1076 | ||
BC012107.1 | - | 8 | 1132 | 4636736 |