Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004786.2 | NP_004777.1 | 6 | 944 | |
NR_024546.1 | 7 | 1049 | |||
XM_006722580.3 | XP_006722643.1 | 7 | 1549 | ||
XM_011526263.2 | XP_011524565.1 | 8 | 1641 | ||
XM_011526264.2 | XP_011524566.1 | 7 | 788 | ||
XM_017026093.1 | XP_016881582.1 | 8 | 1641 | ||
XM_017026094.1 | XP_016881583.1 | 7 | 788 | ||
GenBank mRNA | AB209263.1 | - | 6 | 682 | |
AF003938.1 | - | 6 | 900 | ||
AF051896.1 | - | 6 | 825 | ||
AF052659.1 | - | 6 | 844 | ||
AK094926.1 | - | 8 | 1658 | ||
AK313653.1 | - | 6 | 855 | ||
BC001156.1 | - | 6 | 888 | 3534029 | |
FJ224353.1 | - | 6 | 900 |