Gene Transcripts

Gene Symbol
ITM2B
Entrez Gene ID
9445
Gene Name
integral membrane protein 2B
Gene Aliases
ABRI, BRI, BRI2, BRICD2B, E25B, E3-16, FBD, RDGCA, imBRI2
Chromosome Location
Chr. 13: 48233138 - 48262096 on Build GRCh38
UniGene ID
Hs.643683
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021999.4 NP_068839.1 3 554
GenBank mRNA AF086918.1 - 3 446
AF092128.1 - 3 543
AF136973.1 - 3 543
AF152462.1 - 3 501
AF246221.1 - 3 501
AK000503.1 - 3 486
AK027411.1 - 3 486
AK130048.1 - 3 490
BC000554.2 - 3 480 3163436
BC016148.1 - 3 501 3912066
BT006863.1 - 3 331
DA087198.1 - 3 555

View Full Product Details