Gene Transcripts

Gene Symbol
PSMD6
Entrez Gene ID
9861
Gene Name
proteasome 26S subunit, non-ATPase 6
Gene Aliases
Rpn7, S10, SGA-113M, p42A, p44S10
Chromosome Location
Chr. 3: 64010549 - 64024010 on Build GRCh38
UniGene ID
Hs.152536
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014814.2 NP_055629.1 1, 2 334
GenBank mRNA AF215935.1 - 1, 2 200
AF530062.1 - 1, 2 147
AK290205.1 - 1, 2 188
AY568086.1 - 1, 2 172
BC000630.1 - 1, 2 164 3344973
BC000904.2 - 1, 2 185 2900971
BC012369.1 - 1, 2 182 3858094
D14663.1 - 1, 2 172
HY010377.1 - 1, 2 335

View Full Product Details