Gene Transcripts
- Gene Symbol
- CNKSR1
- Entrez Gene ID
- 10256
- Gene Name
- connector enhancer of kinase suppressor of Ras 1
- Gene Aliases
- CNK, CNK1
- Chromosome Location
- Chr. 1: 26177490 - 26189884 on Build GRCh38
- UniGene ID
- Hs.16232
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001297647.1 |
NP_001284576.1
|
17, 18 |
1613 |
|
NM_001297648.1 |
NP_001284577.1
|
17, 18 |
1587 |
|
NM_006314.2 |
NP_006305.2
|
17, 18 |
1592 |
|
XM_011540484.2 |
XP_011538786.1
|
17, 18 |
1572 |
|
XM_011540485.2 |
XP_011538787.1
|
14, 15 |
1102 |
|
GenBank mRNA |
AB209335.1 |
- |
17, 18 |
1586 |
|
AF100153.1 |
- |
17, 18 |
1570 |
|
AK001900.1 |
- |
17, 18 |
1566 |
|
AK222817.1 |
- |
17, 18 |
1571 |
|
AK222904.1 |
- |
17, 18 |
1575 |
|
AK292646.1 |
- |
17, 18 |
1574 |
|
BC011604.2 |
- |
17, 18 |
1594 |
3139227 |
BC012797.2 |
- |
17, 18 |
1594 |
3505293 |
BT006900.1 |
- |
17, 18 |
1555 |
|
View Full Product Details