Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005870.4 | NP_005861.2 | 2 | 193 | |
GenBank mRNA | AF153608.1 | - | 2 | 131 | |
AK311748.1 | - | 2 | 114 | ||
AY550970.1 | - | 2 | 113 | ||
BC030836.1 | - | 2 | 161 | 4794888 | |
CR457035.1 | - | 2 | 97 | ||
DB171768.1 | - | 2 | 195 | ||
U78303.1 | - | 2 | 128 | ||
U96915.1 | - | 2 | 113 |