Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006387.5 | NP_006378.3 | 7 | 895 | |
GenBank mRNA | AF536542.1 | - | 7 | 862 | |
BC001552.2 | - | 7 | 853 | ||
BC021294.2 | - | 7 | 880 | 4053575 | |
U94836.2 | - | 6 | 797 | ||
Y08265.1 | - | 7 | 852 |