Gene Transcripts
- Gene Symbol
- SRCAP
- Entrez Gene ID
- 10847
- Gene Name
- Snf2-related CREBBP activator protein
- Gene Aliases
- DOMO1, EAF1, FLHS, SWR1
- Chromosome Location
- Chr. 16: 30699141 - 30740129 on Build GRCh38
- UniGene ID
- Hs.620916
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006662.2 |
NP_006653.2
|
34 |
7853 |
|
GenBank mRNA |
AF143946.1 |
- |
29 |
6901 |
|
View Full Product Details