Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001300791.1 | NP_001287720.1 | 7 | 1075 | |
NM_001300792.1 | NP_001287721.1 | 7 | 1075 | ||
NM_007054.6 | NP_008985.3 | 7 | 1075 | ||
XM_006714526.3 | XP_006714589.1 | 7 | 1130 | ||
XM_017008995.1 | XP_016864484.1 | 7 | 1130 | ||
XM_017008996.1 | XP_016864485.1 | 7 | 1130 | ||
GenBank mRNA | AB209780.1 | - | 7 | 1023 | |
AF041853.1 | - | 6 | 916 | ||
AK295089.1 | - | 7 | 1033 | ||
AK313359.1 | - | 7 | 1042 | ||
AM177178.1 | - | 6 | 916 | ||
BC020890.1 | - | 7 | 1040 | ||
BC032599.1 | - | 7 | 1058 | ||
BC039592.1 | - | 7 | 1032 | ||
BC045542.1 | - | 7 | 1069 | 5298675 |