Gene Transcripts

Gene Symbol
TREH
Entrez Gene ID
11181
Gene Name
trehalase
Gene Aliases
TRE, TREA
Chromosome Location
Chr. 11: 118658232 - 118679672 on Build GRCh38
UniGene ID
Hs.129712
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301065.1 NP_001287994.1 7, 8 819
NM_007180.2 NP_009111.2 8, 9 912
GenBank mRNA AB000824.1 - 8, 9 922
AK091981.1 - 6, 7 781
AK223140.1 - 8, 9 886
AK223143.1 - 8, 9 890
BC109206.2 - 7, 8 813 40006501

View Full Product Details