Gene Transcripts

Gene Symbol
CELF3
Entrez Gene ID
11189
Gene Name
CUGBP, Elav-like family member 3
Gene Aliases
BRUNOL1, CAGH4, ERDA4, ETR-1, TNRC4
Chromosome Location
Chr. 1: 151700058 - 151716821 on Build GRCh38
UniGene ID
Hs.26047
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172648.3 NP_001166119.1 1 372
NM_001172649.3 NP_001166120.1 1 876
NM_001291106.1 NP_001278035.1 1 876
NM_001291107.1 NP_001278036.1 1 876
NM_007185.6 NP_009116.3 1 876
XM_006711139.2 XP_006711202.1 2 104
XM_006711141.2 XP_006711204.1 2 104
XM_011509111.2 XP_011507413.1 1 724
GenBank mRNA AB209264.1 - 1 372
AF329264.1 - 1 76
AL519587.3 - 2 108
AY165003.1 - 1 866
BC004145.2 - 2 54
BC041088.1 - 1 108
BC052491.1 - 1 850 4820737
BC104758.1 - 1 139 8143761
BC143226.1 - 1 139 9051727
DR001924.1 - 1 470

View Full Product Details