Gene Transcripts
- Gene Symbol
- LEPROTL1
- Entrez Gene ID
- 23484
- Gene Name
- leptin receptor overlapping transcript-like 1
- Gene Aliases
- HSPC112, Vps55, my047
- Chromosome Location
- Chr. 8: 30095406 - 30137706 on Build GRCh38
- UniGene ID
- Hs.146585
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001128208.1 |
NP_001121680.1
|
2, 3 |
193 |
|
NM_015344.2 |
NP_056159.2
|
2, 3 |
193 |
|
GenBank mRNA |
AF063605.1 |
- |
2, 3 |
170 |
|
AK075363.1 |
- |
2, 3 |
137 |
|
AY277595.1 |
- |
2, 3 |
165 |
|
AY358674.1 |
- |
2, 3 |
165 |
|
BC000642.2 |
- |
2, 3 |
176 |
3344487 |
CR541735.1 |
- |
1, 2 |
86 |
|
CR541760.1 |
- |
1, 2 |
86 |
|
DA258969.1 |
- |
2, 3 |
193 |
|
View Full Product Details