Gene Transcripts
- Gene Symbol
- SAMHD1
- Entrez Gene ID
- 25939
- Gene Name
- SAM and HD domain containing deoxynucleoside triphosphate triphosphohydrolase 1
- Gene Aliases
- CHBL2, DCIP, HDDC1, MOP-5, SBBI88
- Chromosome Location
- Chr. 20: 36890882 - 36951843 on Build GRCh38
- UniGene ID
- Hs.580681
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015474.3 |
NP_056289.2
|
12, 13 |
1619 |
|
XM_005260384.3 |
XP_005260441.1
|
12, 13 |
1592 |
|
XM_011528761.2 |
XP_011527063.1
|
12, 13 |
1592 |
|
GenBank mRNA |
AB013847.1 |
- |
12, 13 |
1419 |
|
AB208944.1 |
- |
12, 13 |
1444 |
|
AF228421.1 |
- |
12, 13 |
1572 |
|
AK027811.1 |
- |
12, 13 |
1592 |
|
AK304187.1 |
- |
12, 13 |
1376 |
|
AK311150.1 |
- |
1, 2 |
54 |
|
AK315169.1 |
- |
12, 13 |
1552 |
|
AL050267.1 |
- |
12, 13 |
1494 |
|
BC036450.2 |
- |
12, 13 |
1447 |
4816438 |
View Full Product Details