Gene Transcripts
- Gene Symbol
- OR2M4
- Entrez Gene ID
- 26245
- Gene Name
- olfactory receptor family 2 subfamily M member 4
- Gene Aliases
- HSHTPCRX18, HTPCRX18, OR1-55, OST710, TPCR100
- Chromosome Location
- Chr. 1: 248238929 - 248239864 on Build GRCh38
- UniGene ID
- Hs.381309
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017504.1 |
NP_059974.1
|
1 |
780 |
|
GenBank mRNA |
X89666.1 |
- |
1 |
406 |
|
View Full Product Details