Gene Transcripts

Gene Symbol
REM1
Entrez Gene ID
28954
Gene Name
RRAD and GEM like GTPase 1
Gene Aliases
GD:REM, GES
Chromosome Location
Chr. 20: 31475273 - 31484905 on Build GRCh38
UniGene ID
Hs.247729
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014012.5 NP_054731.2 2, 3 632
GenBank mRNA AF084465.1 - 1, 2 406
AF152863.1 - 2, 3 614
AK313060.1 - 2, 3 632
BC039813.1 - 2, 3 618 5220518
BT020078.1 - 1, 2 334
BT020079.1 - 1, 2 334

View Full Product Details