Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001083893.1 | NP_001077362.1 | 10 | 1550 | |
NM_014574.3 | NP_055389.3 | 8 | 1298 | ||
XM_005267569.3 | XP_005267626.1 | 9 | 1566 | ||
XM_005267570.3 | XP_005267627.1 | 9 | 1536 | ||
GenBank mRNA | AF243424.1 | - | 9 | 979 | |
AK290229.1 | - | 8 | 1276 | ||
AK314041.1 | - | 8 | 1264 | ||
BC126221.1 | - | 8 | 1216 | 8991937 | |
BC132673.1 | - | 8 | 1216 | 40146695 | |
BM453981.1 | - | 2 | 192 | ||
U17989.1 | - | 8 | 1286 |