Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001270960.1 | NP_001257889.1 | 9 | 928 | |
NM_015953.4 | NP_057037.1 | 10 | 1047 | ||
XM_005258964.4 | XP_005259021.1 | 9 | 964 | ||
XM_006723235.3 | XP_006723298.1 | 9 | 998 | ||
XM_011527015.2 | XP_011525317.1 | 9 | 972 | ||
XM_011527017.2 | XP_011525319.1 | 9 | 1004 | ||
XM_017026851.1 | XP_016882340.1 | 9 | 1022 | ||
XM_017026852.1 | XP_016882341.1 | 9 | 1003 | ||
GenBank mRNA | AF132959.1 | - | 9 | 913 | |
AK092444.1 | - | 7 | 2123 | ||
AK291535.1 | - | 9 | 912 | ||
BC009299.2 | - | 9 | 898 | 4125665 | |
BC010077.2 | - | 10 | 1021 | 4309619 | |
BC011249.1 | - | 8 | 882 | 3452886 |