Gene Transcripts

Gene Symbol
SSH1
Entrez Gene ID
54434
Gene Name
slingshot protein phosphatase 1
Gene Aliases
SSH1L
Chromosome Location
Chr. 12: 108782690 - 108857606 on Build GRCh38
UniGene ID
Hs.199763
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018984.3 NP_061857.3 15 2759
XM_005268984.4 XP_005269041.1 15 3470
XM_005268985.2 XP_005269042.1 14 2585
XM_011538497.1 XP_011536799.1 14 2851
XM_011538499.1 XP_011536801.1 13 2650
XM_011538500.2 XP_011536802.1 14 2675
XM_011538501.2 XP_011536803.1 14 2671
XM_017019491.1 XP_016874980.1 14 3697
XM_017019492.1 XP_016874981.1 15 2812
XM_017019493.1 XP_016874982.1 13 2556
XM_017019494.1 XP_016874983.1 13 2651
GenBank mRNA AB037719.1 - 5 1794
AB072355.1 - 15 2672
BC062341.1 - 15 2751 6376684

View Full Product Details