Gene Transcripts
- Gene Symbol
- DGCR8
- Entrez Gene ID
- 54487
- Gene Name
- DGCR8 microprocessor complex subunit
- Gene Aliases
- C22orf12, DGCRK6, Gy1, pasha
- Chromosome Location
- Chr. 22: 20080232 - 20111877 on Build GRCh38
- UniGene ID
- Hs.643452
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001190326.1 |
NP_001177255.1
|
10, 11 |
2334 |
|
NM_022720.6 |
NP_073557.3
|
11, 12 |
2433 |
|
XM_006724268.2 |
XP_006724331.1
|
8, 9 |
1068 |
|
GenBank mRNA |
AK025539.1 |
- |
10, 11 |
1397 |
|
AK025780.1 |
- |
6, 7 |
709 |
|
AK222796.1 |
- |
10, 11 |
1397 |
|
AY189282.1 |
- |
7, 8 |
727 |
|
BC009323.2 |
- |
9, 10 |
1236 |
|
BC037564.1 |
- |
11, 12 |
2383 |
|
BC078147.1 |
- |
9, 10 |
2048 |
6014346 |
BX649187.1 |
- |
8, 9 |
1325 |
|
CR456356.1 |
- |
10, 11 |
2091 |
|
View Full Product Details