Gene Transcripts

Gene Symbol
VPS13C
Entrez Gene ID
54832
Gene Name
vacuolar protein sorting 13 homolog C
Gene Aliases
PARK23
Chromosome Location
Chr. 15: 61852389 - 62060465 on Build GRCh38
UniGene ID
Hs.511668
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018088.2 NP_001018098.1 71 9852
NM_017684.4 NP_060154.3 69 9723
NM_018080.3 NP_060550.2 69 9723
NM_020821.2 NP_065872.1 71 9852
GenBank mRNA AJ608770.1 - 69 9706
AJ608771.1 - 71 9835
AJ626860.1 - 69 9706
AJ626861.1 - 71 9835
AK123851.1 - 4 410
AK128650.1 - 25 4266
AK304408.1 - 6 757
AL833377.1 - 5 453
CR936716.1 - 20 3334

View Full Product Details