Gene Transcripts

Gene Symbol
PLXNA3
Entrez Gene ID
55558
Gene Name
plexin A3
Gene Aliases
6.3, HSSEXGENE, PLXN3, PLXN4, XAP-6
Chromosome Location
Chr. X: 154458281 - 154473646 on Build GRCh38
UniGene ID
Hs.632839
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017514.4 NP_059984.3 33 5725
XM_005274705.4 XP_005274762.2 33 5665
XM_005274706.4 XP_005274763.2 33 5397
XM_006724829.3 XP_006724892.1 33 5629
XM_011531183.2 XP_011529485.1 32 5089
GenBank mRNA X87852.1 - 33 5734

View Full Product Details