Gene Transcripts
- Gene Symbol
- WDR11
- Entrez Gene ID
- 55717
- Gene Name
- WD repeat domain 11
- Gene Aliases
- BRWD2, DR11, HH14, SRI1, WDR15
- Chromosome Location
- Chr. 10: 120851175 - 120909526 on Build GRCh38
- UniGene ID
- Hs.144447
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018117.11 |
NP_060587.8
|
10, 11 |
1714 |
|
XM_005269962.2 |
XP_005270019.1
|
10, 11 |
1431 |
|
XM_005269963.2 |
XP_005270020.1
|
10, 11 |
1563 |
|
XM_005269964.2 |
XP_005270021.1
|
3, 4 |
469 |
|
XM_017016397.1 |
XP_016871886.1
|
9, 10 |
1402 |
|
XM_017016398.1 |
XP_016871887.1
|
9, 10 |
1332 |
|
XM_017016399.1 |
XP_016871888.1
|
9, 10 |
1276 |
|
XM_017016400.1 |
XP_016871889.1
|
10, 11 |
1605 |
|
GenBank mRNA |
AB037772.2 |
- |
10, 11 |
1527 |
|
AF320223.1 |
- |
10, 11 |
1501 |
|
AK001368.1 |
- |
5, 6 |
617 |
|
AK299524.1 |
- |
3, 4 |
334 |
|
AK315145.1 |
- |
10, 11 |
1492 |
|
AL137699.1 |
- |
8, 9 |
1187 |
|
BC040469.1 |
- |
10, 11 |
1531 |
4730308 |
BC071564.1 |
- |
10, 11 |
1497 |
30346203 |
View Full Product Details