Gene Transcripts

Gene Symbol
TSR1
Entrez Gene ID
55720
Gene Name
TSR1, ribosome maturation factor
Gene Aliases
Chromosome Location
Chr. 17: 2322678 - 2337384 on Build GRCh38
UniGene ID
Hs.388170
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018128.4 NP_060598.3 9, 10 2614
GenBank mRNA AB037822.1 - 9, 10 2737
AK001396.1 - 7, 8 1256
AK026565.1 - 9, 10 1687
BC019090.2 - 8, 9 1522
BC110851.1 - 9, 10 2192 5786818
BC126110.1 - 9, 10 2162 8991826

View Full Product Details