Gene Transcripts

Gene Symbol
PCDHA4
Entrez Gene ID
56144
Gene Name
protocadherin alpha 4
Gene Aliases
CNR1, CNRN1, CRNR1, PCDH-ALPHA4
Chromosome Location
Chr. 5: 140807074 - 141012344 on Build GRCh38
UniGene ID
Hs.199343
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018907.3 NP_061730.1 1 724
NM_031500.2 NP_113688.1 1 724
GenBank mRNA AB208925.1 - 1 724
AF152312.1 - 1 610
AF152482.1 - 1 610
BC112102.1 - 1 696 8327570
BC113609.1 - 1 696 8322661

View Full Product Details