Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020750.2 | NP_065801.1 | 23, 24 | 2881 | |
GenBank mRNA | AB033117.2 | - | 23, 24 | 2820 | |
AF298880.1 | - | 23, 24 | 2733 | ||
AK056619.1 | - | 19, 20 | 2206 | ||
AK291452.1 | - | 23, 24 | 2844 | ||
AL137467.1 | - | 11, 12 | 1324 | ||
BC008347.1 | - | 7, 8 | 890 | ||
BC009969.2 | - | 16, 17 | 1845 | ||
BC062635.1 | - | 23, 24 | 2847 | 5492046 |