Gene Transcripts

Gene Symbol
RBM25
Entrez Gene ID
58517
Gene Name
RNA binding motif protein 25
Gene Aliases
NET52, RED120, RNPC7, S164, Snu71, fSAP94
Chromosome Location
Chr. 14: 73058436 - 73121675 on Build GRCh38
UniGene ID
Hs.531106
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021239.2 NP_067062.1 8, 9 987
XM_011537044.2 XP_011535346.1 9, 10 1089
XM_011537045.2 XP_011535347.1 8, 9 899
GenBank mRNA AK026107.1 - 8, 9 967
AK094697.1 - 7, 8 831
AL832314.1 - 8, 9 958
BC062440.1 - 8, 9 991
BC083496.1 - 8, 9 953
BC111066.1 - 8, 9 951
BC113389.1 - 7, 8 767
BC113391.1 - 7, 8 767
BC136775.1 - 8, 9 824 9020765
BC136776.1 - 8, 9 824 9020766
BC144407.1 - 9, 10 892 9052928
BX647116.1 - 8, 9 940

View Full Product Details