Gene Transcripts

Gene Symbol
ANO3
Entrez Gene ID
63982
Gene Name
anoctamin 3
Gene Aliases
C11orf25, DYT23, DYT24, GENX-3947, TMEM16C
Chromosome Location
Chr. 11: 26189123 - 26663289 on Build GRCh38
UniGene ID
Hs.577269
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001313726.1 NP_001300655.1 6 693
NM_001313727.1 NP_001300656.1 2 251
NM_031418.3 NP_113606.2 5 601
XM_017018118.1 XP_016873607.1 3 229
GenBank mRNA AJ300461.1 - 5 1308
AK295143.1 - 2 164
AK295816.1 - 2 163
AK302659.1 - 6 685
AK304781.1 - 6 534
AK316018.1 - 2 163
BP219022.1 - 2 252
DA247850.1 - 5 561

View Full Product Details