Gene Transcripts

Gene Symbol
GID4
Entrez Gene ID
79018
Gene Name
GID complex subunit 4 homolog
Gene Aliases
C17orf39, VID24
Chromosome Location
Chr. 17: 18039297 - 18068405 on Build GRCh38
UniGene ID
Hs.187422
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024052.4 NP_076957.3 5 980
GenBank mRNA AK127580.1 - 5 667
AK315083.1 - 5 868
BC000636.2 - 5 641
BC041829.1 - 5 985 5268071

View Full Product Details