Gene Transcripts

Gene Symbol
SCRN3
Entrez Gene ID
79634
Gene Name
secernin 3
Gene Aliases
SES3
Chromosome Location
Chr. 2: 174395729 - 174430071 on Build GRCh38
UniGene ID
Hs.470679
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193528.1 NP_001180457.1 2, 3 161
NM_024583.4 NP_078859.2 2, 3 252
XM_005246853.2 XP_005246910.1 2, 3 657
XM_005246854.4 XP_005246911.1 2, 3 1817
XM_005246855.2 XP_005246912.1 2, 3 705
XM_005246856.2 XP_005246913.1 2, 3 661
XM_005246857.2 XP_005246914.1 2, 3 361
XM_017004909.1 XP_016860398.1 2, 3 1249
XM_017004910.1 XP_016860399.1 2, 3 727
GenBank mRNA AK057912.1 - 2, 3 229
AK295357.1 - 2, 3 145
AK296558.1 - 2, 3 251
BC119684.1 - 2, 3 203 40116568
BC119685.1 - 2, 3 203 40116570
BG492502.1 - 2, 3 243
BM725063.1 - 1, 2 243
DA760451.1 - 2, 3 252

View Full Product Details