Gene Transcripts

Gene Symbol
QSER1
Entrez Gene ID
79832
Gene Name
glutamine and serine rich 1
Gene Aliases
Chromosome Location
Chr. 11: 32892826 - 32980270 on Build GRCh38
UniGene ID
Hs.369368
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001076786.2 NP_001070254.1 4 3500
XM_006718325.3 XP_006718388.3 4 3852
XM_017018327.1 XP_016873816.1 4 3852
XM_017018328.1 XP_016873817.1 4 3852
XM_017018329.1 XP_016873818.1 2 4305
XM_017018330.1 XP_016873819.1 3 3250
GenBank mRNA AK025577.1 - 1 227
AK123858.1 - 1 1562
AK125391.1 - 1 2922
AK125902.1 - 1 1048
AK126023.1 - 5 2804
AL834141.1 - 1 2195
BC013746.1 - 1 426
BC112935.1 - 4 3500

View Full Product Details