Gene Transcripts

Gene Symbol
CARD19
Entrez Gene ID
84270
Gene Name
caspase recruitment domain family member 19
Gene Aliases
BinCARD, C9orf89
Chromosome Location
Chr. 9: 93096123 - 93113295 on Build GRCh38
UniGene ID
Hs.434213
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318010.1 NP_001304939.1 2 322
NM_001318011.1 NP_001304940.1 2 322
NM_032310.4 NP_115686.3 2 322
NR_134464.1 2 322
NR_134465.1 2 322
NR_134466.1 2 322
NR_134467.1 2 322
NR_134468.1 2 322
GenBank mRNA AK057716.1 - 2 228
AK091611.1 - 2 228
BC004500.2 - 3 191 3833318
BC038856.1 - 2 215 5215640
BC091476.1 - 2 165
BM930683.1 - 2 167
BX354102.2 - 2 200
CD676329.1 - 2 178
DN998463.1 - 2 215
HY005780.1 - 2 323

View Full Product Details