Gene Transcripts

Gene Symbol
L3MBTL3
Entrez Gene ID
84456
Gene Name
l(3)mbt-like 3 (Drosophila)
Gene Aliases
MBT-1, MBT1
Chromosome Location
Chr. 6: 130018583 - 130141449 on Build GRCh38
UniGene ID
Hs.658051
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007102.2 NP_001007103.1 20, 21 2247
NM_032438.2 NP_115814.1 21, 22 2322
XM_005267161.4 XP_005267218.1 21, 22 2498
XM_006715576.3 XP_006715639.1 21, 22 2427
XM_006715578.3 XP_006715641.1 21, 22 2306
XM_011536179.2 XP_011534481.1 21, 22 2305
XM_011536180.2 XP_011534482.1 21, 22 2324
XM_011536181.2 XP_011534483.1 21, 22 2259
XM_011536183.2 XP_011534485.1 21, 22 2855
XM_011536184.2 XP_011534486.1 21, 22 2259
XM_017011355.1 XP_016866844.1 20, 21 2338
XM_017011356.1 XP_016866845.1 20, 21 2230
GenBank mRNA AB058701.1 - 20, 21 2220
AK092756.1 - 11, 12 1499
AK094137.1 - 21, 22 2329
AK290219.1 - 20, 21 2239
BC060845.1 - 20, 21 2241 30342272

View Full Product Details