Gene Transcripts

Gene Symbol
ACSS1
Entrez Gene ID
84532
Gene Name
acyl-CoA synthetase short-chain family member 1
Gene Aliases
ACAS2L, ACECS1, AceCS2L
Chromosome Location
Chr. 20: 25006230 - 25058182 on Build GRCh38
UniGene ID
Hs.529353
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252675.1 NP_001239604.1 7 1263
NM_001252676.1 NP_001239605.1 6 1090
NM_001252677.1 NP_001239606.1 7 1263
NM_032501.3 NP_115890.2 7 1263
XM_011529388.1 XP_011527690.1 6 1048
GenBank mRNA AB058749.1 - 2 180
AK024396.1 - 1 2297
AK024424.1 - 1 1824
AK027817.1 - 1 86
AK092295.1 - 5 672
AK125058.1 - 5 2466
AK127566.1 - 7 1209
AK296306.1 - 6 1087
AK314440.1 - 7 1186
BC039261.1 - 7 1222 5298849
BC044588.1 - 7 1188 5260496
BC055008.1 - 7 1188 4838976
BC114612.1 - 7 1054

View Full Product Details