Gene Transcripts

Gene Symbol
SLITRK2
Entrez Gene ID
84631
Gene Name
SLIT and NTRK like family member 2
Gene Aliases
CXorf2, SLITL1
Chromosome Location
Chr. X: 145817829 - 145829856 on Build GRCh38
UniGene ID
Hs.320368
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144003.2 NP_001137475.1 5 2633
NM_001144004.2 NP_001137476.1 5 2621
NM_001144005.2 NP_001137477.1 5 2643
NM_001144006.2 NP_001137478.1 3 2853
NM_001144008.2 NP_001137480.1 3 2865
NM_001144009.2 NP_001137481.1 2 2706
NM_001144010.2 NP_001137482.1 3 2843
NM_032539.4 NP_115928.1 5 3116
XM_005262342.3 XP_005262399.1 5 3718
XM_005262343.3 XP_005262400.1 5 3728
XM_005262344.3 XP_005262401.1 5 3706
XM_005262345.3 XP_005262402.1 4 3569
GenBank mRNA AK091015.1 - 1 2234
AK096278.1 - 5 3112
AK289732.1 - 5 2483
BC113011.2 - 3 2707 40028691
BC113012.2 - 3 2709 40028693
CR627412.1 - 2 4155

View Full Product Details