Gene Transcripts

Gene Symbol
C8orf76
Entrez Gene ID
84933
Gene Name
chromosome 8 open reading frame 76
Gene Aliases
Chromosome Location
Chr. 8: 123219956 - 123241398 on Build GRCh38
UniGene ID
Hs.707401
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032847.2 NP_116236.1 4 714
GenBank mRNA AK027731.1 - 4 693
AK222660.1 - 4 662
BC067796.1 - 4 688 5260773
DR156993.1 - 4 714

Gene Symbol
ZHX1-C8orf76
Entrez Gene ID
100533106
Gene Name
ZHX1-C8orf76 readthrough
Gene Aliases
Chromosome Location
Chr. 8: 123226189 - 123274487 on Build GRCh38
UniGene ID
Hs.707401
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204180.1 NP_001191109.1 5 930
GenBank mRNA BC012379.2 - 4 660

View Full Product Details