Gene Transcripts

Gene Symbol
SSH2
Entrez Gene ID
85464
Gene Name
slingshot protein phosphatase 2
Gene Aliases
SSH-2, SSH-2L
Chromosome Location
Chr. 17: 29625938 - 29930239 on Build GRCh38
UniGene ID
Hs.654754
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282129.1 NP_001269058.1 9 972
NM_033389.3 NP_203747.2 8 703
XM_005258058.3 XP_005258115.1 10 865
XM_005258059.2 XP_005258116.1 7 839
XM_006722149.3 XP_006722212.1 7 4470
XM_011525402.2 XP_011523704.1 9 700
XM_011525404.2 XP_011523706.1 7 1463
XM_011525405.2 XP_011523707.1 7 1511
XM_011525406.2 XP_011523708.1 6 790
XM_011525407.1 XP_011523709.1 6 535
XM_017025251.1 XP_016880740.1 9 908
XM_017025252.1 XP_016880741.1 7 769
XM_017025253.1 XP_016880742.1 9 911
XM_017025254.1 XP_016880743.1 9 911
GenBank mRNA AB072358.1 - 8 663
AB099290.1 - 8 663
AK296413.1 - 9 837

View Full Product Details