Gene Transcripts

Gene Symbol
PNPT1
Entrez Gene ID
87178
Gene Name
polyribonucleotide nucleotidyltransferase 1
Gene Aliases
COXPD13, DFNB70, OLD35, PNPASE, old-35
Chromosome Location
Chr. 2: 55634063 - 55693910 on Build GRCh38
UniGene ID
Hs.388733
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033109.4 NP_149100.2 7, 8 663
XM_005264629.2 XP_005264686.1 7, 8 593
XM_017005172.1 XP_016860661.1 6, 7 543
GenBank mRNA AJ458465.1 - 7, 8 594
AK307589.1 - 593
AY027528.1 - 7, 8 590
BC053660.1 - 7, 8 577 6062060

View Full Product Details