Gene Transcripts

Gene Symbol
SFXN5
Entrez Gene ID
94097
Gene Name
sideroflexin 5
Gene Aliases
BBG-TCC
Chromosome Location
Chr. 2: 72942036 - 73072234 on Build GRCh38
UniGene ID
Hs.368171
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144579.2 NP_653180.1 3 333
XM_005264648.1 XP_005264705.1 3 239
XM_006712147.1 XP_006712210.1 3 239
XM_011533186.2 XP_011531488.1 4 1881
XM_011533187.2 XP_011531489.1 4 1878
XM_011533188.2 XP_011531490.1 3 1736
XM_011533189.1 XP_011531491.1 3 239
XM_011533190.2 XP_011531492.1 4 786
XM_011533192.2 XP_011531494.1 4 1020
XM_011533194.2 XP_011531496.1 4 1879
XM_011533195.2 XP_011531497.1 4 1881
XM_011533196.2 XP_011531498.1 3 965
XM_011533197.1 XP_011531499.1 3 239
XM_017005340.1 XP_016860829.1 3 1733
XM_017005341.1 XP_016860830.1 3 1734
XM_017005342.1 XP_016860831.1 3 239
XM_017005343.1 XP_016860832.1 4 1020
XM_017005344.1 XP_016860833.1 3 965
XM_017005346.1 XP_016860835.1 4 261
GenBank mRNA AK024193.1 - 2 861
AK289731.1 - 3 222
AK294751.1 - 3 270
AK295637.1 - 3 144
AK314770.1 - 3 221
AL833454.1 - 4 262
AY044437.1 - 3 337
BC013233.1 - 3 274
BC101311.2 - 3 333 40024811
BC101312.1 - 3 333 40024812
BC101313.2 - 3 333 40024813

View Full Product Details