Gene Transcripts
- Gene Symbol
- SLC5A11
- Entrez Gene ID
- 115584
- Gene Name
- solute carrier family 5 member 11
- Gene Aliases
- KST1, RKST1, SGLT6, SMIT2
- Chromosome Location
- Chr. 16: 24832784 - 24911711 on Build GRCh38
- UniGene ID
- Hs.164118
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001258413.2 |
NP_001245342.1
|
5, 6 |
604 |
|
NM_001258414.1 |
NP_001245343.1
|
5, 6 |
604 |
|
NM_052944.4 |
NP_443176.2
|
5, 6 |
614 |
|
XM_005255080.2 |
XP_005255137.1
|
6, 7 |
642 |
|
XM_017022900.1 |
XP_016878389.1
|
6, 7 |
958 |
|
XM_017022901.1 |
XP_016878390.1
|
6, 7 |
1668 |
|
XM_017022902.1 |
XP_016878391.1
|
6, 7 |
797 |
|
XM_017022903.1 |
XP_016878392.1
|
7, 8 |
807 |
|
XM_017022904.1 |
XP_016878393.1
|
7, 8 |
977 |
|
XM_017022906.1 |
XP_016878395.1
|
6, 7 |
818 |
|
XM_017022907.1 |
XP_016878396.1
|
6, 7 |
642 |
|
XM_017022908.1 |
XP_016878397.1
|
6, 7 |
701 |
|
GenBank mRNA |
AF292385.1 |
- |
4, 5 |
382 |
|
AJ305237.1 |
- |
4, 5 |
400 |
|
AK125267.1 |
- |
7, 8 |
730 |
|
AK298373.1 |
- |
4, 5 |
399 |
|
AY044906.1 |
- |
5, 6 |
614 |
|
BC049385.1 |
- |
5, 6 |
474 |
5755223 |
BC057780.1 |
- |
5, 6 |
473 |
5287366 |
U41898.1 |
- |
2, 3 |
166 |
|
View Full Product Details