Gene Transcripts

Gene Symbol
CYYR1
Entrez Gene ID
116159
Gene Name
cysteine and tyrosine rich 1
Gene Aliases
C21orf95
Chromosome Location
Chr. 21: 26466209 - 26573404 on Build GRCh38
UniGene ID
Hs.37445
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320768.1 NP_001307697.1 2 607
NM_052954.3 NP_443186.2 2 607
NR_135472.1 2 607
XM_011529450.2 XP_011527752.1 2 487
GenBank mRNA AF401639.1 - 2 212
AK054581.1 - 2 487
AK223576.1 - 2 324
AK304124.1 - 2 487
AK313231.1 - 2 393
AL833200.1 - 2 418
AY061853.1 - 2 152
BC036761.1 - 2 395 5588436
JF693912.1 - 2 155
JF693913.1 - 2 212
JF693914.1 - 1 80
KC843341.1 - 2 202
KF154022.1 - 2 212

View Full Product Details