Gene Transcripts

Gene Symbol
OR2D3
Entrez Gene ID
120775
Gene Name
olfactory receptor family 2 subfamily D member 3
Gene Aliases
OR11-89
Chromosome Location
Chr. 11: 6921002 - 6921994 on Build GRCh38
UniGene ID
Hs.446766
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004684.1 NP_001004684.1 1 345
GenBank mRNA BC137208.1 - 1 373
BC137209.1 - 1 373 9021206

View Full Product Details